missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. p-7133611
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N009025ML 25 mL
N0090100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N009025ML Supplier TCI America Supplier No. N009025ML
Only null left

Chemical Identifiers

CAS 500-22-1
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006382
InChI Key QJZUKDFHGGYHMC-UHFFFAOYSA-N
Synonym 3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde
PubChem CID 10371
ChEBI CHEBI:28345
IUPAC Name pyridine-3-carbaldehyde
SMILES C1=CC(=CN=C1)C=O

Specifications

Color Yellow
Boiling Point 97°C
Quantity 25 mL
UN Number 1989
Formula Weight 107.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pyridinecarboxaldehyde
EINECSNumber (5)-6320
TSCA Yes