accessibility menu, dialog, popup

UserName

CAS RN 500-22-1

IUPAC Name: pyridine-3-carbaldehyde
Synonyms: 3-pyridinecarboxaldehyde 3-pyridylcarboxaldehyde nicotinealdehyde rowalind
Molecular Weight (g/mol): 107.11
Molecular Formula: C6H5NO
InChi Key: QJZUKDFHGGYHMC-UHFFFAOYSA-N
SMILES: O=CC1=CC=CN=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)