missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinecarboxaldehyde, 98%

Catalog No. p-4509898
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131830250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC131830250 Supplier Thermo Scientific Chemicals Supplier No. 131830250
Only null left

CAS: 500-22-1 | C6H5NO | 107.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 500-22-1
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.11
InChI Key QJZUKDFHGGYHMC-UHFFFAOYSA-N
Synonym 3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde
PubChem CID 10371
ChEBI CHEBI:28345
IUPAC Name pyridine-3-carbaldehyde
SMILES C1=CC(=CN=C1)C=O

Specifications

Color Yellow to Brown
Density 1.1420g/mL
Boiling Point 95.0°C to 97.0°C (15.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5480 to 1.5500
Linear Formula (C5H4N)COH
Quantity 25 g
Beilstein 21,I,288
Specific Gravity 1.142
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 107.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Pyridinecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
EINECSNumber 207-900-4

RUO – Research Use Only