missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyridin-3-ylaniline, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 57976-57-5 |
|---|---|
| Molecular Formula | C11H10N2 |
| Molecular Weight (g/mol) | 170.215 |
| InChI Key | YTJQJGKMRLQBJP-UHFFFAOYSA-N |
| Synonym | 3-pyridin-3-yl benzenamine, 3-pyridin-3-yl aniline, 3-3-aminophenyl pyridine, 3-3-pyridyl aniline, benzenamine, 3-3-pyridinyl, 3-3-pyridinyl aniline, 3-pyridin-3-yl anilin, 3-3-pyridyl phenylamine, 3-pyridine-3-yl aniline, 3-pyridin-3-yl-phenylamine |
| PubChem CID | 459521 |
| IUPAC Name | 3-pyridin-3-ylaniline |
| SMILES | C1=CC(=CC(=C1)N)C2=CN=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC39814CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC39814DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 57976-57-5 | |
| 170.215 | |
| 3-pyridin-3-yl benzenamine, 3-pyridin-3-yl aniline, 3-3-aminophenyl pyridine, 3-3-pyridyl aniline, benzenamine, 3-3-pyridinyl, 3-3-pyridinyl aniline, 3-pyridin-3-yl anilin, 3-3-pyridyl phenylamine, 3-pyridine-3-yl aniline, 3-pyridin-3-yl-phenylamine | |
| 3-pyridin-3-ylaniline |
| C11H10N2 | |
| YTJQJGKMRLQBJP-UHFFFAOYSA-N | |
| 459521 | |
| C1=CC(=CC(=C1)N)C2=CN=CC=C2 |
Specifications
| 57976-57-5 | |
| C11H10N2 | |
| YTJQJGKMRLQBJP-UHFFFAOYSA-N | |
| 3-pyridin-3-ylaniline | |
| 459521 | |
| 97% |
| 97% | |
| 3-pyridin-3-yl benzenamine, 3-pyridin-3-yl aniline, 3-3-aminophenyl pyridine, 3-3-pyridyl aniline, benzenamine, 3-3-pyridinyl, 3-3-pyridinyl aniline, 3-pyridin-3-yl anilin, 3-3-pyridyl phenylamine, 3-pyridine-3-yl aniline, 3-pyridin-3-yl-phenylamine | |
| C1=CC(=CC(=C1)N)C2=CN=CC=C2 | |
| 170.215 | |
| 170.21 | |
| 3-Pyridin-3-ylaniline |