accessibility menu, dialog, popup

UserName

CAS RN 57976-57-5

NH2N

CAS RN 57976-57-5

IUPAC Name: 3-(pyridin-3-yl)aniline
Synonyms: 3-(pyridin-3-yl)aniline
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: YTJQJGKMRLQBJP-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)