missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridin-3-ylaniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC39814DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC39814DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

57976-57-5
170.215
3-pyridin-3-yl benzenamine, 3-pyridin-3-yl aniline, 3-3-aminophenyl pyridine, 3-3-pyridyl aniline, benzenamine, 3-3-pyridinyl, 3-3-pyridinyl aniline, 3-pyridin-3-yl anilin, 3-3-pyridyl phenylamine, 3-pyridine-3-yl aniline, 3-pyridin-3-yl-phenylamine
3-pyridin-3-ylaniline
C11H10N2
YTJQJGKMRLQBJP-UHFFFAOYSA-N
459521
C1=CC(=CC(=C1)N)C2=CN=CC=C2

Specifications

3-Pyridin-3-ylaniline
97%
C11H10N2
3-pyridin-3-yl benzenamine, 3-pyridin-3-yl aniline, 3-3-aminophenyl pyridine, 3-3-pyridyl aniline, benzenamine, 3-3-pyridinyl, 3-3-pyridinyl aniline, 3-pyridin-3-yl anilin, 3-3-pyridyl phenylamine, 3-pyridine-3-yl aniline, 3-pyridin-3-yl-phenylamine
C1=CC(=CC(=C1)N)C2=CN=CC=C2
170.215
170.21
57976-57-5
Amber Glass Bottle
1g
YTJQJGKMRLQBJP-UHFFFAOYSA-N
3-pyridin-3-ylaniline
459521
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions