missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Propyltoluene 99.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 181°C |
| Quantity | 5 mL |
| UN Number | 3295 |
| Formula Weight | 134.22 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Propyltoluene |
Safety and Handling
| EINECSNumber | (3)-0015 |
| TSCA | Yes |