missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Propionylpyridine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 77°C |
| Quantity | 5 g |
| Formula Weight | 135.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Propionylpyridine |
Safety and Handling
| TSCA | No |