accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 1570-48-5

IUPAC Name: 1-(pyridin-3-yl)propan-1-one
Synonyms: 1-(pyridin-3-yl)propan-1-one
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: VDNKJMUNLKAGAM-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results