missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 898289-01-5
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD09025831
InChI Key QIWMAMIMWHCYGZ-UHFFFAOYSA-N
Synonym 3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl
PubChem CID 18525731
IUPAC Name 3-piperidin-1-ylpyridine-2-carboxylic acid
SMILES C1CCN(CC1)C2=C(N=CC=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC37601CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC37601DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

898289-01-5
206.245
QIWMAMIMWHCYGZ-UHFFFAOYSA-N
18525731
C1CCN(CC1)C2=C(N=CC=C2)C(=O)O
C11H14N2O2
MFCD09025831
3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl
3-piperidin-1-ylpyridine-2-carboxylic acid

Specifications

898289-01-5
100.0
C11H14N2O2
3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl
C1CCN(CC1)C2=C(N=CC=C2)C(=O)O
206.245
206.24
3-Piperidinopyridine-2-carboxylic acid
96.5
97%
MFCD09025831
QIWMAMIMWHCYGZ-UHFFFAOYSA-N
3-piperidin-1-ylpyridine-2-carboxylic acid
18525731
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents