missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC37601CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC37601CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

898289-01-5
206.245
QIWMAMIMWHCYGZ-UHFFFAOYSA-N
18525731
C1CCN(CC1)C2=C(N=CC=C2)C(=O)O
C11H14N2O2
MFCD09025831
3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl
3-piperidin-1-ylpyridine-2-carboxylic acid

Specifications

3-Piperidinopyridine-2-carboxylic acid
96.5
97%
C11H14N2O2
MFCD09025831
QIWMAMIMWHCYGZ-UHFFFAOYSA-N
3-piperidin-1-ylpyridine-2-carboxylic acid
18525731
97%
898289-01-5
100.0
Amber Glass Bottle
250mg
3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl
C1CCN(CC1)C2=C(N=CC=C2)C(=O)O
206.245
206.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents