missing translation for 'onlineSavingsMsg'
Learn More

(3-Piperidinophenyl)methanol, 97%, Thermo Scientific™

$242.71 - $242.71

Chemical Identifiers

CAS 852180-58-6
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD07772816
InChI Key HNESBVGXCDQHLJ-UHFFFAOYSA-N
Synonym 3-piperidinophenyl methanol, 3-piperidin-1-yl phenyl methanol, 3-piperidin-1-ylphenyl methanol, 3-piperidylphenyl methan-1-ol, benzenemethanol,3-1-piperidinyl
PubChem CID 7162330
IUPAC Name (3-piperidin-1-ylphenyl)methanol
SMILES C1CCN(CC1)C2=CC=CC(=C2)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC34009DA
SDS View Documents
Thermo Scientific Chemicals
CC34009DA
1 g Amber Glass Bottle
Each for $242.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

852180-58-6
191.274
HNESBVGXCDQHLJ-UHFFFAOYSA-N
7162330
C1CCN(CC1)C2=CC=CC(=C2)CO
C12H17NO
MFCD07772816
3-piperidinophenyl methanol, 3-piperidin-1-yl phenyl methanol, 3-piperidin-1-ylphenyl methanol, 3-piperidylphenyl methan-1-ol, benzenemethanol,3-1-piperidinyl
(3-piperidin-1-ylphenyl)methanol

Specifications

852180-58-6
100.0
Amber Glass Bottle
MFCD07772816
3-piperidinophenyl methanol, 3-piperidin-1-yl phenyl methanol, 3-piperidin-1-ylphenyl methanol, 3-piperidylphenyl methan-1-ol, benzenemethanol,3-1-piperidinyl
C1CCN(CC1)C2=CC=CC(=C2)CO
191.274
191.27
(3-Piperidinophenyl)methanol
96.5
97%
C12H17NO
1 g
HNESBVGXCDQHLJ-UHFFFAOYSA-N
(3-piperidin-1-ylphenyl)methanol
7162330
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'