missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidinomethyl)benzaldehyde, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC45204DE

Quantity 5g

 View more versions of this product

Catalog No. CC45204DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

471930-01-5
203.285
UJDTVJKIOWAUNQ-UHFFFAOYSA-N
7164643
C1CCN(CC1)CC2=CC=CC(=C2)C=O
C13H17NO
MFCD07782418
3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
3-(piperidin-1-ylmethyl)benzaldehyde

Specifications

3-(Piperidinomethyl)benzaldehyde
96.5
97%
5g
3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
C1CCN(CC1)CC2=CC=CC(=C2)C=O
203.285
203.28
471930-01-5
100.0
C13H17NO
MFCD07782418
UJDTVJKIOWAUNQ-UHFFFAOYSA-N
3-(piperidin-1-ylmethyl)benzaldehyde
7164643
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents