missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidinomethyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC45204DA
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45204DA 1g
CC45204DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC45204DA Supplier Thermo Scientific Chemicals Supplier No. CC45204DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 471930-01-5
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD07782418
InChI Key UJDTVJKIOWAUNQ-UHFFFAOYSA-N
Synonym 3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
PubChem CID 7164643
IUPAC Name 3-(piperidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(CC1)CC2=CC=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 203.28
Percent Purity 97%
Chemical Name or Material 3-(Piperidinomethyl)benzaldehyde
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.