missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Piperidinecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America N0420500G
Specifications
| 3-Piperidinecarboxylic Acid | |
| Yellow | |
| MFCD00005992 MFCD01630787 | |
| nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine | |
| OC(=O)C1CCCNC1 | |
| 129.16 | |
| CHEBI:116931 | |
| ≥98.0% (T) |
| 498-95-3 | |
| C6H11NO2 | |
| 500 g | |
| XJLSEXAGTJCILF-UHFFFAOYSA-N | |
| piperidine-3-carboxylic acid | |
| 4498 | |
| 129.16 | |
| Crystalline Powder |
Chemical Identifiers
| 498-95-3 | |
| 129.16 | |
| XJLSEXAGTJCILF-UHFFFAOYSA-N | |
| 4498 | |
| piperidine-3-carboxylic acid |
| C6H11NO2 | |
| MFCD00005992 MFCD01630787 | |
| nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine | |
| CHEBI:116931 | |
| OC(=O)C1CCCNC1 |
Safety and Handling
RTECSNumber : TM6125380
TSCA : No