missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133801
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N042025G 25 g
N0420500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N042025G Supplier TCI America Supplier No. N042025G
Only null left

Chemical Identifiers

CAS 498-95-3
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00005992 MFCD01630787
InChI Key XJLSEXAGTJCILF-UHFFFAOYSA-N
Synonym nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine
PubChem CID 4498
ChEBI CHEBI:116931
IUPAC Name piperidine-3-carboxylic acid
SMILES OC(=O)C1CCCNC1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Piperidinecarboxylic Acid
RTECSNumber TM6125380
TSCA No