missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline, 99%

CAS: 108-99-6 | C6H7N | 93.129 g/mol

Supplier:  Thermo Scientific Chemicals A14012AE

 View more versions of this product

Catalog No. AAA14012AE


Only null left
Explore available promotions

Description

Description

3-Picoline is used as a precursor in pharmaceuticals and agricultural industries. It acts as a precursor to 3-cyanopyridine, niacin, vitamin-B. It is an antidote for organophosphate poisoning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Picoline is used as a precursor in pharmaceuticals and agricultural industries. It acts as a precursor to 3-cyanopyridine, niacin, vitamin-B. It is an antidote for organophosphate poisoning.

Solubility
Miscible with water, alcohol, ether, acetone, carbon tetrachloride.

Notes
Hygroscopic. Incompatible with acids, acid chlorides and chloroformates.
Specifications

Specifications

3-Picoline
-18°C
143°C to 144°C
C6H7N
MFCD00006402
UN2313
Hygroscopic
3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine
ITQTTZVARXURQS-UHFFFAOYSA-N
3-methylpyridine
7970
93.13
108-99-6
0.955
36°C (96°F)
1.5055
100 mL
1366
14,7401
Miscible with water,alcohol,ether,acetone,carbon tetrachloride.
CC1=CN=CC=C1
93.129
CHEBI:39922
99%

Chemical Identifiers

108-99-6
93.129
ITQTTZVARXURQS-UHFFFAOYSA-N
7970
3-methylpyridine
C6H7N
MFCD00006402
3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine
CHEBI:39922
CC1=CN=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H332-H311-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES

EINECSNumber : 203-636-9

RTECSNumber : TJ5000000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only