missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline, 99%

Catalog No. p-7021609
Change view
Click to view available options
Quantity:
100 mL
2500 mL
Catalog No. Quantity
AAA14012AE 100 mL
AAA140120F 2500 mL
2 options

Catalog No. AAA14012AE

Supplier: Thermo Scientific Chemicals A14012AE

Only null left

CAS: 108-99-6 | C6H7N | 93.129 g/mol

3-Picoline is used as a precursor in pharmaceuticals and agricultural industries. It acts as a precursor to 3-cyanopyridine, niacin, vitamin-B. It is an antidote for organophosphate poisoning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Picoline is used as a precursor in pharmaceuticals and agricultural industries. It acts as a precursor to 3-cyanopyridine, niacin, vitamin-B. It is an antidote for organophosphate poisoning.

Solubility
Miscible with water, alcohol, ether, acetone, carbon tetrachloride.

Notes
Hygroscopic. Incompatible with acids, acid chlorides and chloroformates.

Chemical Identifiers

CAS 108-99-6
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006402
InChI Key ITQTTZVARXURQS-UHFFFAOYSA-N
Synonym 3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine
PubChem CID 7970
ChEBI CHEBI:39922
IUPAC Name 3-methylpyridine
SMILES CC1=CN=CC=C1

Specifications

Melting Point -18°C
Density 0.955
Boiling Point 143°C to 144°C
Flash Point 36°C (96°F)
Refractive Index 1.5055
Quantity 100 mL
UN Number UN2313
Beilstein 1366
Sensitivity Hygroscopic
Merck Index 14,7401
Solubility Information Miscible with water,alcohol,ether,acetone,carbon tetrachloride.
Formula Weight 93.13
Percent Purity 99%
Chemical Name or Material 3-Picoline
Show More Show Less
Hazard Category H226-H302+H332-H311-H314
Hazard Statement GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES
EINECSNumber 203-636-9
RTECSNumber TJ5000000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only