missing translation for 'onlineSavingsMsg'
Learn More

3-Phthalimidopropionic Acid 98.0+%, TCI America™

Catalog No. p-7135407
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21015G 5 g
P210125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21015G Supplier TCI America Supplier No. P21015G
Only null left

Chemical Identifiers

CAS 3339-73-9
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00023096
InChI Key DXXHRZUOTPMGEH-UHFFFAOYSA-N
Synonym 3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
PubChem CID 76859
IUPAC Name 3-(1,3-dioxoisoindol-2-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O

Specifications

Melting Point 153°C
Color White
Quantity 5 g
Formula Weight 219.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phthalimidopropionic Acid