missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phosphoshikimic Acid lithium salt, 95.0% (HPLC), MilliporeSigma™ Supelco™
$904.11 - $904.11
Chemical Identifiers
| CAS | 63959-45-5 |
|---|---|
| Molecular Formula | C7H11O8P |
| Molecular Weight (g/mol) | 254.13 |
| MDL Number | MFCD04039676 |
| InChI Key | QYOJSKGCWNAKGW-PBXRRBTRSA-N |
| Synonym | Lithium 3-phosphoshikimate; Lithium shikimate-3-phosphate; S-3-P; S3P; Shikimic acid-3-phosphate lithium salt |
| IUPAC Name | (3R,4S,5R)-4,5-dihydroxy-3-(phosphonooxy)cyclohex-1-ene-1-carboxylic acid |
| SMILES | O[C@@H]1CC(=C[C@@H](OP(O)(O)=O)[C@H]1O)C(O)=O |
Chemical Identifiers
| 63959-45-5 | |
| 254.13 | |
| QYOJSKGCWNAKGW-PBXRRBTRSA-N | |
| (3R,4S,5R)-4,5-dihydroxy-3-(phosphonooxy)cyclohex-1-ene-1-carboxylic acid |
| C7H11O8P | |
| MFCD04039676 | |
| Lithium 3-phosphoshikimate; Lithium shikimate-3-phosphate; S-3-P; S3P; Shikimic acid-3-phosphate lithium salt | |
| O[C@@H]1CC(=C[C@@H](OP(O)(O)=O)[C@H]1O)C(O)=O |
Specifications
| 63959-45-5 | |
| C7H11O8P · xLi+ | |
| 10 mg | |
| QYOJSKGCWNAKGW-PBXRRBTRSA-N | |
| (3R,4S,5R)-4,5-dihydroxy-3-(phosphonooxy)cyclohex-1-ene-1-carboxylic acid | |
| 254.13 (free acid basis) |
| C7H11O8P | |
| MFCD04039676 | |
| Lithium 3-phosphoshikimate; Lithium shikimate-3-phosphate; S-3-P; S3P; Shikimic acid-3-phosphate lithium salt | |
| O[C@@H]1CC(=C[C@@H](OP(O)(O)=O)[C@H]1O)C(O)=O | |
| 254.13 | |
| ≥95.0% (HPLC) |