missing translation for 'onlineSavingsMsg'
Learn More

3-Phosphonopropionic Acid 98.0+%, TCI America™

Catalog No. P23485G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23485G 5 g
P23481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23485G Supplier TCI America Supplier No. P23485G
Only null left

Chemical Identifiers

CAS 5962-42-5
Molecular Formula C3H7O5P
Molecular Weight (g/mol) 154.058
MDL Number MFCD00002770
InChI Key NLBSQHGCGGFVJW-UHFFFAOYSA-N
Synonym 3-phosphonopropionic acid, propanoic acid, 3-phosphono, 2-carboxyethylphosphonic acid, beta-phosphonopropionic acid, 1iig, acmc-20amyo, 3-phosphopropionic acid, 3-phosphono-propanoicaci, 3-phosphonoproprionic acid, ambscpod_20/0652
PubChem CID 1682
IUPAC Name 3-phosphonopropanoic acid
SMILES C(CP(=O)(O)O)C(=O)O

Specifications

Melting Point 168°C
Quantity 5 g
UN Number 3261
Formula Weight 154.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phosphonopropionic Acid
RTECSNumber UA2558437
TSCA Yes