accessibility menu, dialog, popup

UserName

HOOPOHOHO

CAS RN 5962-42-5

IUPAC Name: 3-phosphonopropanoic acid
Synonyms: 2-carboxyethylphosphonic acid
Molecular Weight (g/mol): 154.06
Molecular Formula: C3H7O5P
InChi Key: NLBSQHGCGGFVJW-UHFFFAOYSA-N
SMILES: OC(=O)CCP(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results