missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyltoluene, 95%, Thermo Scientific™

$129.97 - $501.34

Chemical Identifiers

CAS 643-93-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00008533
InChI Key NPDIDUXTRAITDE-UHFFFAOYSA-N
Synonym 3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
PubChem CID 12564
IUPAC Name 1-methyl-3-phenylbenzene
SMILES CC1=CC=CC(=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC309840010
SDS View Documents
Thermo Scientific Chemicals
309840010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC309840050
SDS View Documents
Thermo Scientific Chemicals
309840050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

643-93-6
168.24
NPDIDUXTRAITDE-UHFFFAOYSA-N
12564
CC1=CC=CC(=C1)C2=CC=CC=C2
C13H12
MFCD00008533
3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
1-methyl-3-phenylbenzene

Specifications

643-93-6
100.0
272°C
Authentic
Glass bottle
1.6010 to 1.605
MFCD00008533
1.01
NPDIDUXTRAITDE-UHFFFAOYSA-N
1-methyl-3-phenylbenzene
12564
95%
94.0
1.0100g/mL
>110°C
94% min. (GC)
C13H12
C6H5C6H4CH3
1g
3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
CC1=CC=CC(=C1)C2=CC=CC=C2
168.24
168.24
3-Phenyltoluene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-404-3

SDS
Documents

Documents

RUO – Research Use Only