accessibility menu, dialog, popup

UserName

CH3

CAS RN 643-93-6

IUPAC Name: 3-methyl-1,1'-biphenyl
Synonyms: 3-methylbiphenyl
Molecular Weight (g/mol): 168.24
Molecular Formula: C13H12
InChi Key: NPDIDUXTRAITDE-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (7)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (5)