missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyltoluene, 95%

Catalog No. AC309840010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309840010 1 g Glass bottle
AC309840050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC309840010 Supplier Thermo Scientific Chemicals Supplier No. 309840010

CAS: 643-93-6 | C13H12 | 168.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 643-93-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00008533
InChI Key NPDIDUXTRAITDE-UHFFFAOYSA-N
Synonym 3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
PubChem CID 12564
IUPAC Name 1-methyl-3-phenylbenzene
SMILES CC1=CC=CC(=C1)C2=CC=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 4°C to 5°C
Color Yellow
Density 1.0100g/mL
Boiling Point 272°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.6010 to 1.605
Linear Formula C6H5C6H4CH3
Quantity 1 g
Specific Gravity 1.01
Formula Weight 168.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Phenyltoluene
Show More Show Less
EINECSNumber 211-404-3

RUO – Research Use Only