missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylthiophene 95.0+%, TCI America™
$193.86 - $659.87
Chemical Identifiers
| CAS | 2404-87-7 |
|---|---|
| Molecular Formula | C10H8S |
| Molecular Weight (g/mol) | 160.23 |
| MDL Number | MFCD00114997 |
| InChI Key | ZDQZVKVIYAPRON-UHFFFAOYSA-N |
| PubChem CID | 75473 |
| IUPAC Name | 3-phenylthiophene |
| SMILES | S1C=CC(=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 2404-87-7 | |
| 160.23 | |
| ZDQZVKVIYAPRON-UHFFFAOYSA-N | |
| 3-phenylthiophene |
| C10H8S | |
| MFCD00114997 | |
| 75473 | |
| S1C=CC(=C1)C1=CC=CC=C1 |
Specifications
| 2404-87-7 | |
| White-Yellow | |
| C10H8S | |
| 1 g | |
| S1C=CC(=C1)C1=CC=CC=C1 | |
| 160.23 | |
| 160.23 | |
| Crystalline Powder |
| 91°C | |
| 256°C | |
| MFCD00114997 | |
| ZDQZVKVIYAPRON-UHFFFAOYSA-N | |
| 3-phenylthiophene | |
| 75473 | |
| ≥95.0% (GC) | |
| 3-Phenylthiophene |
Safety and Handling
TSCA : No