accessibility menu, dialog, popup

UserName

CAS RN 2404-87-7

S

CAS RN 2404-87-7

IUPAC Name: 3-phenylthiophene
Synonyms: 3-phenylthiophene
Molecular Weight (g/mol): 160.23
Molecular Formula: C10H8S
InChi Key: ZDQZVKVIYAPRON-UHFFFAOYSA-N
SMILES: S1C=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results