missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylthiophene 95.0+%, TCI America™

Catalog No. P11265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11265G 5 g
P11261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11265G Supplier TCI America Supplier No. P11265G
Only null left

Chemical Identifiers

CAS 2404-87-7
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.23
MDL Number MFCD00114997
InChI Key ZDQZVKVIYAPRON-UHFFFAOYSA-N
PubChem CID 75473
IUPAC Name 3-phenylthiophene
SMILES S1C=CC(=C1)C1=CC=CC=C1

Specifications

Melting Point 91°C
Color White-Yellow
Boiling Point 256°C
Quantity 5 g
Formula Weight 160.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylthiophene