missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylthiophene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 91°C |
| Color | White-Yellow |
| Boiling Point | 256°C |
| Quantity | 1 g |
| Formula Weight | 160.23 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Phenylthiophene |
Safety and Handling
| TSCA | No |