Learn More
3-(Phenylsulfonyl)propionitrile, 99%
CAS: 10154-75-3 | C9H9NO2S | 195.236 g/mol
Supplier: Thermo Scientific Chemicals L2019606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-(Phenylsulfonyl)propionitrile | |
| 94°C to 96°C | |
| C9H9NO2S | |
| 5 g | |
| 3-phenylsulfonyl propionitrile, 3-phenylsulfonyl propanenitrile, 3-phenylsulphonyl propiononitrile, 3-benzenesulfonyl propanenitrile, 3-phenylsulfonylpropionitrile, acmc-1bvrh, enamine_005271, 3-phenylsulfonyl propanitrile, 3-phenylsulphonyl-propionitrile, propanenitrile, 3-phenylsulfonyl | |
| C1=CC=C(C=C1)S(=O)(=O)CCC#N | |
| 195.236 | |
| 195.24 |
| 10154-75-3 | |
| Odorless | |
| MFCD00047798 | |
| 2051019 | |
| OVZPZPVSUAKTCM-UHFFFAOYSA-N | |
| 3-(benzenesulfonyl)propanenitrile | |
| 82413 | |
| 99% |
Chemical Identifiers
| 10154-75-3 | |
| 195.236 | |
| OVZPZPVSUAKTCM-UHFFFAOYSA-N | |
| 82413 | |
| C1=CC=C(C=C1)S(=O)(=O)CCC#N |
| C9H9NO2S | |
| MFCD00047798 | |
| 3-phenylsulfonyl propionitrile, 3-phenylsulfonyl propanenitrile, 3-phenylsulphonyl propiononitrile, 3-benzenesulfonyl propanenitrile, 3-phenylsulfonylpropionitrile, acmc-1bvrh, enamine_005271, 3-phenylsulfonyl propanitrile, 3-phenylsulphonyl-propionitrile, propanenitrile, 3-phenylsulfonyl | |
| 3-(benzenesulfonyl)propanenitrile |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 233-424-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only