missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpyrazole 98.0+%, TCI America™
$175.22 - $521.94
Chemical Identifiers
| CAS | 2458-26-6 |
|---|---|
| Molecular Formula | C9H8N2 |
| Molecular Weight (g/mol) | 144.18 |
| MDL Number | MFCD00159654 |
| InChI Key | OEDUIFSDODUDRK-UHFFFAOYSA-N |
| Synonym | 3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole |
| PubChem CID | 17155 |
| IUPAC Name | 5-phenyl-1H-pyrazole |
| SMILES | N1N=CC=C1C1=CC=CC=C1 |
Chemical Identifiers
| 2458-26-6 | |
| 144.18 | |
| OEDUIFSDODUDRK-UHFFFAOYSA-N | |
| 17155 | |
| N1N=CC=C1C1=CC=CC=C1 |
| C9H8N2 | |
| MFCD00159654 | |
| 3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole | |
| 5-phenyl-1H-pyrazole |
Specifications
| 2458-26-6 | |
| 314°C | |
| MFCD00159654 | |
| 3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole | |
| N1N=CC=C1C1=CC=CC=C1 | |
| 144.18 | |
| 144.18 | |
| Crystalline Powder |
| 78°C | |
| C9H8N2 | |
| 1 g | |
| OEDUIFSDODUDRK-UHFFFAOYSA-N | |
| 5-phenyl-1H-pyrazole | |
| 17155 | |
| ≥98.0% (GC,T) | |
| 3-Phenylpyrazole |
Safety and Handling
RTECSNumber : UQ7950000
TSCA : No