accessibility menu, dialog, popup

UserName

NHN

CAS RN 2458-26-6

IUPAC Name: 5-phenyl-1H-pyrazole
Synonyms: 3-phenyl-2H-pyrazole
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: OEDUIFSDODUDRK-UHFFFAOYSA-N
SMILES: N1N=CC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)