missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpyrazole 98.0+%, TCI America™

Catalog No. P20631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20631G 1 g
P20635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20631G Supplier TCI America Supplier No. P20631G

Chemical Identifiers

CAS 2458-26-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00159654
InChI Key OEDUIFSDODUDRK-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
PubChem CID 17155
IUPAC Name 5-phenyl-1H-pyrazole
SMILES N1N=CC=C1C1=CC=CC=C1

Specifications

Melting Point 78°C
Boiling Point 314°C
Quantity 1 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylpyrazole
RTECSNumber UQ7950000
TSCA No