missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpyrazole-5-carboxylic Acid Hydrate 97.0+%, TCI America™

Supplier:  TCI America P20211G

 View more versions of this product

Catalog No. P20211G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Phenylpyrazole-5-carboxylic Acid Hydrate
233°C
C10H7N2O2
1 g
[O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1
187.18
188.19
Crystalline Powder
5071-61-4
White-Yellow
MFCD01248821,MFCD05170017
QBPUOAJBMXXBNU-UHFFFAOYSA-M
3-phenyl-1H-pyrazole-5-carboxylate
121025
≥97.0% (HPLC,T)

Chemical Identifiers

5071-61-4
187.18
QBPUOAJBMXXBNU-UHFFFAOYSA-M
3-phenyl-1H-pyrazole-5-carboxylate
C10H7N2O2
MFCD01248821,MFCD05170017
121025
[O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UQ6437000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions