missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpyrazole-5-carboxylic Acid Hydrate 97.0+%, TCI America™
$305.70 - $305.70
Chemical Identifiers
| CAS | 5071-61-4 |
|---|---|
| Molecular Formula | C10H7N2O2 |
| Molecular Weight (g/mol) | 187.18 |
| MDL Number | MFCD01248821,MFCD05170017 |
| InChI Key | QBPUOAJBMXXBNU-UHFFFAOYSA-M |
| PubChem CID | 121025 |
| IUPAC Name | 3-phenyl-1H-pyrazole-5-carboxylate |
| SMILES | [O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1 |
Chemical Identifiers
| 5071-61-4 | |
| 187.18 | |
| QBPUOAJBMXXBNU-UHFFFAOYSA-M | |
| 3-phenyl-1H-pyrazole-5-carboxylate |
| C10H7N2O2 | |
| MFCD01248821,MFCD05170017 | |
| 121025 | |
| [O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1 |
Specifications
| 5071-61-4 | |
| White-Yellow | |
| MFCD01248821,MFCD05170017 | |
| QBPUOAJBMXXBNU-UHFFFAOYSA-M | |
| 3-phenyl-1H-pyrazole-5-carboxylate | |
| 121025 | |
| ≥97.0% (HPLC,T) | |
| 3-Phenylpyrazole-5-carboxylic Acid Hydrate |
| 233°C | |
| C10H7N2O2 | |
| 1 g | |
| [O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1 | |
| 187.18 | |
| 188.19 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UQ6437000
TSCA : No