missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58569AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58569AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58569AD Supplier Thermo Scientific Chemicals Supplier No. H58569AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626207-42-9 | C9H11BrZn | 264.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 626207-42-9
Molecular Formula C9H11BrZn
Molecular Weight (g/mol) 264.471
MDL Number MFCD08064069
InChI Key XTDLRSRUPYQIGX-UHFFFAOYSA-M
Synonym 3-phenyl-1-propylzinc bromide, bromo 3-phenylpropyl zinc, 3-phenyl-1-propylzinc bromide 0.5 m in tetrahydrofuran, 3-phenylpropylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 24722663
IUPAC Name bromozinc(1+);propylbenzene
SMILES [CH2-]CCC1=CC=CC=C1.[Zn+]Br

Specifications

Linear Formula C6H5CH2CH2CH2ZnBr
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Formula Weight 264.47
Chemical Name or Material 3-Phenylpropylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only