accessibility menu, dialog, popup

UserName

CAS RN 626207-42-9

IUPAC Name: 3-phenylpropan-1-ide; bromozincylium
Synonyms: 3-phenylpropan-1-ide; bromozincylium
Molecular Weight (g/mol): 264.47
Molecular Formula: C9H11BrZn
InChi Key: XTDLRSRUPYQIGX-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results