missing translation for 'onlineSavingsMsg'
Learn More

(3-Phenylpropyl)triphenylphosphonium bromide, 97+%

Catalog No. p-7020639
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1266909 10 g
AAA1266918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1266909 Supplier Thermo Scientific Chemicals Supplier No. A1266909

CAS: 7484-37-9 | C27H26BrP | 461.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7484-37-9
Molecular Formula C27H26BrP
Molecular Weight (g/mol) 461.38
MDL Number MFCD00051886
InChI Key RPUZOJFXAPSSJD-UHFFFAOYSA-M
Synonym 3-phenylpropyl triphenylphosphonium bromide, triphenyl 3-phenylpropyl phosphonium bromide, triphenyl 3-phenylpropyl phosphanium bromide, 3-phenylpropyltriphenylphosphoniumbromide, phenylpropyl triphenylphosphonium bromide, 3-phenylpropyltriphenylphosphonium bromide, triphenyl 3-phenylpropylphosphonium bromide, 3-phenyl-propyl-triphenylphosphonium bromide, 3-phenylpropyl triphenyl phosphonium bromide
PubChem CID 10928647
IUPAC Name triphenyl(3-phenylpropyl)phosphanium;bromide
SMILES [Br-].C(CC1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 208°C to 212°C
Odor Odorless
Linear Formula C6H5(CH2)2P(C6H5)3Br
Quantity 10 g
Beilstein 4173336
Sensitivity Hygroscopic
Formula Weight 461.39
Percent Purity 98%
Chemical Name or Material (3-Phenylpropyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-289-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only