missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropyl Isobutyrate 98.0+%, TCI America™

Catalog No. i042525ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I042525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I042525ML Supplier TCI America Supplier No. I042525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-58-2
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00082227
InChI Key VBTAKMZSMFMLGT-UHFFFAOYSA-N
Synonym Isobutyric Acid 3-Phenylpropyl Ester
PubChem CID 7662
IUPAC Name 3-phenylpropyl 2-methylpropanoate
SMILES CC(C)C(=O)OCCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 137°C
Quantity 25 mL
Formula Weight 206.29
Physical Form Liquid
Chemical Name or Material 3-Phenylpropyl Isobutyrate
EINECSNumber (3)-1041&(3)-1045
RTECSNumber NQ5440000
TSCA Yes