accessibility menu, dialog, popup

UserName

CAS RN 103-58-2

IUPAC Name: 3-phenylpropyl 2-methylpropanoate
Synonyms: hydrocinnamyl isobutyrate
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: VBTAKMZSMFMLGT-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results