missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropyl Bromide 97.0+%, TCI America™

Catalog No. B1237500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1237500G 500 g
B123725G 25 g
B1237100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1237500G Supplier TCI America Supplier No. B1237500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-59-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000257
InChI Key XMZQWZJMTBCUFT-UHFFFAOYSA-N
Synonym 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
PubChem CID 12503
IUPAC Name (3-bromopropyl)benzene
SMILES BrCCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 129°C
Quantity 500 g
Formula Weight 199.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropyl Bromide