missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionyl Chloride 97.0+%, TCI America™

Catalog No. H069625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H069625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H069625G Supplier TCI America Supplier No. H069625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 645-45-4
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000748
InChI Key MFEILWXBDBCWKF-UHFFFAOYSA-N
Synonym 3-phenylpropionyl chloride, hydrocinnamoyl chloride, benzenepropanoyl chloride, dihydrocinnamoyl chloride, hydrocinnamyl chloride, propionyl chloride, 3-phenyl, hydrocinnamoylchloride, beta-phenylpropionyl chloride, 3-phenylpropionic acid chloride, beta-phenylpropanoyl chloride
PubChem CID 64801
IUPAC Name 3-phenylpropanoyl chloride
SMILES C1=CC=C(C=C1)CCC(=O)Cl

Specifications

Color Yellow
Boiling Point 225°C
Quantity 25 g
UN Number 3265
Formula Weight 168.62
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropionyl Chloride
RTECSNumber MW5652395
TSCA Yes