accessibility menu, dialog, popup

UserName

CAS RN 645-45-4

IUPAC Name: 3-phenylpropanoyl chloride
Synonyms: benzenepropanoyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: MFEILWXBDBCWKF-UHFFFAOYSA-N
SMILES: ClC(=O)CCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)