missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionyl chloride, 98%

Catalog No. AAA1086030
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1086030 250 g
AAA1086009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1086030 Supplier Thermo Scientific Chemicals Supplier No. A1086030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 645-45-4 | C9H9ClO | 168.62 g/mol

The 3-phenylpropionate (dihydrocinnamoyl) ester has been used as a protecting group in nucleoside synthesis. Used in the synthesis of 1-Phenyl-3-methyl-4-acyl-5- pyrazolones. N-(3-phenyl-n-propyl)-1-phenyl-2-aminopropane were prepared by acylation of the appropriate amine with 3-phenylpropionyl chloride followed by reduction of the resulting amide with either LiA1H4. For the preparation of the phenylacetyl (PA) and 3-phenylpropionyl (PP) derivatives, the corresponding amino acids were coupled in the usual manner with phenylacetyl chloride or 3-phenylpropionyl chloride in the presence of either NaOH(PA-LD- and DL-Pgly. 3-Phenylpropionyl chloride (4) was treated with diazomethane to give diazoketone. Allyloxy resin 12 was acylated with 3-phenylpropionyl chloride and treated with 50% TFA for 1 h followed by treatment with Pd(PPh3)4/PPh3 in solution to furnish the desired hydrox- amic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The 3-phenylpropionate (dihydrocinnamoyl) ester has been used as a protecting group in nucleoside synthesis. Used in the synthesis of 1-Phenyl-3-methyl-4-acyl-5- pyrazolones. N-(3-phenyl-n-propyl)-1-phenyl-2-aminopropane were prepared by acylation of the appropriate amine with 3-phenylpropionyl chloride followed by reduction of the resulting amide with either LiA1H4. For the preparation of the phenylacetyl (PA) and 3-phenylpropionyl (PP) derivatives, the corresponding amino acids were coupled in the usual manner with phenylacetyl chloride or 3-phenylpropionyl chloride in the presence of either NaOH(PA-LD- and DL-Pgly. 3-Phenylpropionyl chloride (4) was treated with diazomethane to give diazoketone. Allyloxy resin 12 was acylated with 3-phenylpropionyl chloride and treated with 50% TFA for 1 h followed by treatment with Pd(PPh3)4/PPh3 in solution to furnish the desired hydrox- amic acid.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, amines, alcohols, active metals, bases, water, moisture.

Chemical Identifiers

CAS 645-45-4
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000748
InChI Key MFEILWXBDBCWKF-UHFFFAOYSA-N
Synonym 3-phenylpropionyl chloride, hydrocinnamoyl chloride, benzenepropanoyl chloride, dihydrocinnamoyl chloride, hydrocinnamyl chloride, propionyl chloride, 3-phenyl, hydrocinnamoylchloride, beta-phenylpropionyl chloride, 3-phenylpropionic acid chloride, beta-phenylpropanoyl chloride
PubChem CID 64801
IUPAC Name 3-phenylpropanoyl chloride
SMILES C1=CC=C(C=C1)CCC(=O)Cl

Specifications

Melting Point -7°C
Density 1.135
Boiling Point 225°C
Flash Point 108°C (226°F)
Odor Acrid
Refractive Index 1.5265
Quantity 250 g
UN Number UN3265
Beilstein 742586
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts),and ether.
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 3-Phenylpropionyl chloride
Show More Show Less
Hazard Category H314-H331-H335
Hazard Statement GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 211-443-6
RTECSNumber MW5652395
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only