missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionitrile, 98%

Catalog No. p-7044447
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0648309 10 g
AAL0648318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0648309 Supplier Thermo Scientific Chemicals Supplier No. L0648309
Only null left

CAS: 645-59-0 | C9H9N | 131.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-59-0
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001961
InChI Key ACRWYXSKEHUQDB-UHFFFAOYSA-N
Synonym 3-phenylpropionitrile, benzenepropanenitrile, hydrocinnamonitrile, 2-phenylethyl cyanide, benzenepropionitrile, 2-cyanoethyl benzene, 3-phenylpropiononitrile, phenylpropionitrile, phenethyl cyanide, hydrocinnamique nitrile
PubChem CID 12581
ChEBI CHEBI:85426
IUPAC Name 3-phenylpropanenitrile
SMILES C1=CC=C(C=C1)CCC#N

Specifications

Melting Point -2°C
Density 1.008
Boiling Point 261°C
Flash Point >110°C (230°F)
Refractive Index 1.521
Quantity 10 g
UN Number UN3276
Beilstein 636348
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 3-Phenylpropionitrile
Show More Show Less
Hazard Category H301-H311-H331
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 211-447-8
RTECSNumber MW5604750
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only