missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionamide 98.0+%, TCI America™

Supplier:  TCI America P184525G

 View more versions of this product

Catalog No. P184525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Phenylpropionamide
101°C
C9H11NO
25 g
C1=CC=C(C=C1)CCC(=O)N
149.193
149.19
Crystalline Powder
102-93-2
White-Yellow
MFCD00025535
VYIBCOSBNVFEIW-UHFFFAOYSA-N
3-phenylpropanamide
7625
≥98.0% (GC,N)

Chemical Identifiers

102-93-2
149.193
VYIBCOSBNVFEIW-UHFFFAOYSA-N
3-phenylpropanamide
C9H11NO
MFCD00025535
7625
C1=CC=C(C=C1)CCC(=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MW4905500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions