missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpropionamide 98.0+%, TCI America™
$152.08 - $448.56
Chemical Identifiers
| CAS | 102-93-2 |
|---|---|
| Molecular Formula | C9H11NO |
| Molecular Weight (g/mol) | 149.193 |
| MDL Number | MFCD00025535 |
| InChI Key | VYIBCOSBNVFEIW-UHFFFAOYSA-N |
| PubChem CID | 7625 |
| IUPAC Name | 3-phenylpropanamide |
| SMILES | C1=CC=C(C=C1)CCC(=O)N |
Chemical Identifiers
| 102-93-2 | |
| 149.193 | |
| VYIBCOSBNVFEIW-UHFFFAOYSA-N | |
| 3-phenylpropanamide |
| C9H11NO | |
| MFCD00025535 | |
| 7625 | |
| C1=CC=C(C=C1)CCC(=O)N |
Specifications
| 102-93-2 | |
| White-Yellow | |
| MFCD00025535 | |
| VYIBCOSBNVFEIW-UHFFFAOYSA-N | |
| 3-phenylpropanamide | |
| 7625 | |
| ≥98.0% (GC,N) | |
| 3-Phenylpropionamide |
| 101°C | |
| C9H11NO | |
| 5 g | |
| C1=CC=C(C=C1)CCC(=O)N | |
| 149.193 | |
| 149.19 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : MW4905500
TSCA : No