missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionaldehyde 93.0+%, TCI America™

Catalog No. p021725g
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P021725G 25 g
P0217100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P021725G Supplier TCI America Supplier No. P021725G
Only null left

Chemical Identifiers

CAS 104-53-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00007021
InChI Key YGCZTXZTJXYWCO-UHFFFAOYSA-N
Synonym benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl
PubChem CID 7707
IUPAC Name 3-phenylpropanal
SMILES C1=CC=C(C=C1)CCC=O

Specifications

Color Yellow
Boiling Point 223°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropionaldehyde
EINECSNumber (3)-2656
RTECSNumber MW4890000
TSCA Yes