accessibility menu, dialog, popup

UserName

O

CAS RN 104-53-0

IUPAC Name: 3-phenylpropanal
Synonyms: benzenepropanal
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: YGCZTXZTJXYWCO-UHFFFAOYSA-N
SMILES: O=CCCC1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)