missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpropionaldehyde 93.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 223°C |
| Quantity | 25 g |
| Formula Weight | 134.18 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Phenylpropionaldehyde |
Safety and Handling
| EINECSNumber | (3)-2656 |
| RTECSNumber | MW4890000 |
| TSCA | Yes |